Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: DDX24 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_020414.3 | NP_065147.1 | 5 | 1892 | ||
GenBank mRNA | AB209382.1 | - | 4 | 1812 | ||
AF134475.1 | - | 3 | 708 | |||
AF145022.1 | - | 3 | 708 | |||
AF214731.1 | - | 5 | 1891 | |||
AK025162.1 | - | 5 | 1888 | |||
AK294613.1 | - | 3 | 730 | |||
AK296726.1 | - | 6 | 1603 | |||
AK297232.1 | - | 5 | 1756 | |||
AK304504.1 | - | 4 | 1159 | |||
AL136886.1 | - | 5 | 1868 | |||
BC008847.2 | - | 5 | 1868 | 4301025 | ||
BC009406.2 | - | 5 | 1868 | 4303448 | ||
BC096826.1 | - | 6 | 1698 | 30925011 | ||
BX537533.1 | - | 5 | 1879 |