Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: KNL1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_144508.4 | NP_653091.3 | 11, 12 | 5682 | ||
NM_170589.4 | NP_733468.3 | 12, 13 | 5760 | |||
XM_017022432.1 | XP_016877921.1 | 10, 11 | 5885 | |||
GenBank mRNA | AB046790.1 | - | 2, 3 | 2654 | ||
AF173994.1 | - | 12, 13 | 5989 | |||
AF461041.1 | - | 11, 12 | 5666 |