Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
Integrated CDMO-CRO Services
CDMO Services
CRO Services
Custom Services
Enterprise Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Thermo Fisher Scientific
Search All
Search
Search button
Search button
Close
Order Status
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Services Central
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other PCNP SIRNA Assays ›
Gene Symbol:
PCNP
Gene Name:
PEST proteolytic signal containing nuclear protein
Gene Aliases:
-
Chromosome Location:
Chr.3: 101574094 - 101594465 on Build GRCh38
UniGene ID:
Hs.732633
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID
s32653
Purification
HPLC
Standard
HPLC-IVR
Size
1 nmol
5 nmol
InVivo | 5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
1 nmol
5 nmol
InVivo | 5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
1 nmol
5 nmol
InVivo | 5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
Availability
Inventoried
Catalog #
4392420
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: PCNP
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_001320395.1
NP_001307324.1
4
534
NM_001320397.1
NP_001307326.1
5
491
NM_001320398.1
NP_001307327.1
5
587
NM_001320399.1
NP_001307328.1
6
716
NM_001320400.1
NP_001307329.1
3
459
NM_001320401.1
NP_001307330.1
4
515
NM_020357.2
NP_065090.1
5
590
NR_135223.1
5
538
NR_135224.1
4
375
NR_135225.1
5
535
NR_135226.1
3
407
NR_135227.1
5
791
NR_135228.1
4
482
NR_135229.1
5
719
NR_135230.1
4
463
NR_135231.1
3
300
NR_135232.1
6
633
XM_017006922.1
XP_016862411.1
4
445
GenBank mRNA
AB037675.1
-
5
486
AK025551.1
-
5
430
AK098760.1
-
5
470
AK222978.1
-
6
481
AK314629.1
-
5
471
BC013916.1
-
4
442
3929908
BC022001.2
-
5
488
4823990
BC107796.1
-
5
432
BF244508.1
-
349
BI547939.1
-
5
703
BI596913.1
-
6
642
BI913146.1
-
5
615
BX647886.1
-
1
1174
CN482876.1
-
6
517
CR627371.1
-
4
276
DA041324.1
-
4
390
DA304502.1
-
5
538
DA710370.1
-
4
405
DA817951.1
-
5
455
DA821754.1
-
3
322
DB032930.1
-
4
314
DB060329.1
-
5
402
DB481581.1
-
4
351
HY051831.1
-
3
188
Back To Top
More Information
Gene Ontology Categories:
Function(s)
Process(es)
cell cycle
protein ubiquitination
proteasome-mediated ubiquitin-dependent protein catabolic process
protein binding
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-blue-5c9c5c4749-vpmkj:80/100.66.78.144:80.
git-commit: d22414ba47b22cc02ffc24003f0e179da61511a6
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.21.0-Offline