Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
Integrated CDMO-CRO Services
CDMO Services
CRO Services
Custom Services
Enterprise Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Thermo Fisher Scientific
Search All
Search
Search button
Search button
Close
Order Status
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Services Central
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other PCNP SIRNA Assays ›
Gene Symbol:
PCNP
Gene Name:
PEST proteolytic signal containing nuclear protein
Gene Aliases:
-
Chromosome Location:
Chr.3: 101574094 - 101594465 on Build GRCh38
UniGene ID:
Hs.732633
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID
s32654
Purification
HPLC
Standard
HPLC-IVR
Size
1 nmol
5 nmol
InVivo | 5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
1 nmol
5 nmol
InVivo | 5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
1 nmol
5 nmol
InVivo | 5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
Availability
Inventoried
Catalog #
4392420
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: PCNP
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_001320395.1
NP_001307324.1
4
560
NM_001320397.1
NP_001307326.1
5
517
NM_001320398.1
NP_001307327.1
5
613
NM_001320399.1
NP_001307328.1
6
742
NM_001320400.1
NP_001307329.1
3
485
NM_001320401.1
NP_001307330.1
4
541
NM_020357.2
NP_065090.1
5
616
NR_135223.1
5
564
NR_135224.1
4
401
NR_135225.1
5
561
NR_135226.1
3
433
NR_135227.1
5
817
NR_135228.1
4
508
NR_135229.1
5
745
NR_135230.1
4
489
NR_135231.1
3
326
NR_135232.1
6
659
XM_017006922.1
XP_016862411.1
4
471
GenBank mRNA
AB037675.1
-
5
512
AK025551.1
-
5
456
AK098760.1
-
5
496
AK222978.1
-
6
507
AK314629.1
-
5
497
BC013916.1
-
4
468
3929908
BC022001.2
-
5
514
4823990
BC107796.1
-
5
458
BF244508.1
-
375
BF541732.1
-
5
507
BI913146.1
-
5
641
CN482876.1
-
6
543
CR627371.1
-
4
302
DA041324.1
-
4
416
DA304502.1
-
5
564
DA710370.1
-
4
431
DA817951.1
-
5
481
DA821754.1
-
3
348
DB032930.1
-
4
340
DB060329.1
-
5
428
DB481581.1
-
4
377
HY051831.1
-
3
214
Back To Top
More Information
Other Species Targeted
This siRNA also targets the following transcripts in Mouse: NM_001024622, AK159201, BC145194, BC023320, BC106192, and others.
Gene Ontology Categories:
Function(s)
Process(es)
cell cycle
protein ubiquitination
proteasome-mediated ubiquitin-dependent protein catabolic process
protein binding
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-blue-5c9c5c4749-ffltp:80/100.66.78.226:80.
git-commit: d22414ba47b22cc02ffc24003f0e179da61511a6
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.21.0-Offline