Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PARP11 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_020367.5 | NP_065100.2 | 3 | 404 | ||
NR_104461.1 | 3 | 404 | ||||
XM_005253711.4 | XP_005253768.1 | 3 | 798 | |||
XM_011520970.1 | XP_011519272.1 | 4 | 520 | |||
XM_011520971.2 | XP_011519273.1 | 4 | 920 | |||
XM_011520973.2 | XP_011519275.1 | 3 | 233 | |||
XM_017019670.1 | XP_016875159.1 | 4 | 442 | |||
GenBank mRNA | AF263540.1 | - | 2 | 219 | ||
AK314916.1 | - | 3 | 305 | |||
BC017569.1 | - | 3 | 330 | 4077842 | ||
BC031073.1 | - | 3 | 392 | 5284275 | ||
BX953946.1 | - | 3 | 404 |