Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TIGAR | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_020375.2 | NP_065108.1 | 4 | 327 | ||
GenBank mRNA | AJ272206.1 | - | 4 | 315 | ||
AK313226.1 | - | 4 | 300 | |||
AY425618.1 | - | 4 | 286 | |||
BC012340.1 | - | 4 | 286 | 4576725 |