Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PNPLA2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_020376.3 | NP_065109.1 | 10 | 1963 | ||
XM_006718265.3 | XP_006718328.1 | 9 | 2111 | |||
XM_006718266.3 | XP_006718329.1 | 8 | 2051 | |||
XM_017018028.1 | XP_016873517.1 | 9 | 1909 | |||
GenBank mRNA | AF055000.1 | - | 8 | 1481 | ||
AL833707.1 | - | 4 | 1190 | |||
AY203925.1 | - | 6 | 2614 | |||
BC011958.1 | - | 7 | 1347 | |||
BC017280.2 | - | 10 | 1962 | 4875483 | ||
HQ651812.1 | - | 10 | 1963 | |||
JF279441.1 | - | 10 | 1963 | |||
JF279442.1 | - | 10 | 1963 | |||
KF601369.1 | - | 10 | 1963 |