Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: REXO4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001279349.1 | NP_001266278.1 | 3 | 722 | ||
NM_001279350.1 | NP_001266279.1 | 5 | 1060 | |||
NM_001279351.1 | NP_001266280.1 | 5 | 921 | |||
NM_020385.3 | NP_065118.2 | 5 | 1238 | |||
NR_103995.1 | 3 | 697 | ||||
NR_103996.1 | 4 | 891 | ||||
GenBank mRNA | AF273304.1 | - | 5 | 1186 | ||
AF295774.1 | - | 5 | 1186 | |||
AK000247.1 | - | 2 | 99 | |||
AK025493.1 | - | 5 | 1092 | |||
AK295982.1 | - | 5 | 921 | |||
AK304547.1 | - | 5 | 976 | |||
AK310164.1 | - | 3 | 600 | |||
AK314333.1 | - | 5 | 1152 | |||
AK315904.1 | - | 6 | 911 | |||
AL136894.1 | - | 5 | 1169 | |||
BC009274.2 | - | 5 | 1141 | 3958331 | ||
BC071691.1 | - | 4 | 750 | |||
BX333016.2 | - | 3 | 612 |