Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: REXO4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001279349.1 | NP_001266278.1 | 3, 4 | 742 | ||
NM_001279350.1 | NP_001266279.1 | 5, 6 | 1080 | |||
NM_001279351.1 | NP_001266280.1 | 5, 6 | 941 | |||
NM_020385.3 | NP_065118.2 | 5, 6 | 1258 | |||
NR_103995.1 | 3, 4 | 717 | ||||
NR_103996.1 | 4, 5 | 911 | ||||
GenBank mRNA | AF273304.1 | - | 5, 6 | 1206 | ||
AF295774.1 | - | 5, 6 | 1206 | |||
AK000247.1 | - | 2, 3 | 119 | |||
AK025493.1 | - | 5, 6 | 1112 | |||
AK295982.1 | - | 5, 6 | 941 | |||
AK304547.1 | - | 5, 6 | 996 | |||
AK310164.1 | - | 3, 4 | 620 | |||
AK314333.1 | - | 5, 6 | 1172 | |||
AK315904.1 | - | 6, 7 | 931 | |||
AL136894.1 | - | 5, 6 | 1189 | |||
BC009274.2 | - | 5, 6 | 1161 | 3958331 | ||
BC071691.1 | - | 4, 5 | 770 | |||
BX333016.2 | - | 3, 4 | 632 |