Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PGLYRP4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_020393.3 | NP_065126.2 | 6 | 874 | ||
XM_011509789.2 | XP_011508091.1 | 6 | 886 | |||
XM_011509790.1 | XP_011508092.1 | 6 | 874 | |||
XM_011509791.2 | XP_011508093.1 | 6 | 865 | |||
XM_011509792.1 | XP_011508094.1 | 6 | 874 | |||
XM_011509793.2 | XP_011508095.1 | 3 | 562 | |||
GenBank mRNA | AF242518.1 | - | 6 | 590 | ||
AK292203.1 | - | 6 | 877 | |||
AY035377.1 | - | 6 | 579 | |||
BC107157.1 | - | 5 | 561 | 40032748 | ||
BC107158.1 | - | 5 | 561 | 40032750 | ||
BC142636.1 | - | 6 | 849 | 40147772 |