Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: LPAR5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001142961.1 | NP_001136433.1 | 2 | 593 | ||
NM_020400.5 | NP_065133.1 | 2 | 753 | |||
GenBank mRNA | AB041943.1 | - | 1 | 570 | ||
AJ272207.1 | - | 1 | 312 | |||
AK313127.1 | - | 2 | 567 | |||
BC033571.2 | - | 2 | 544 | 4297800 | ||
BC072394.1 | - | 2 | 743 | 5751923 |
Set Membership: |
G-protein coupled receptor |