Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: NUP107 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_020401.2 | NP_065134.1 | 27 | 2718 | ||
XM_005269037.3 | XP_005269094.1 | 26 | 2649 | |||
XM_011538576.1 | XP_011536878.1 | 27 | 2726 | |||
GenBank mRNA | AJ295745.1 | - | 27 | 2718 | ||
AK002079.1 | - | 17 | 1670 | |||
AK302773.1 | - | 27 | 2760 | |||
AK315157.1 | - | 27 | 2682 | |||
BC017167.2 | - | 24 | 2254 | 3909848 | ||
BC043343.2 | - | 27 | 2643 | 5785959 |