Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PLXDC1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_020405.4 | NP_065138.2 | 6 | 910 | ||
GenBank mRNA | AF279144.2 | - | 6 | 780 | ||
AF378753.1 | - | 6 | 780 | |||
AK093589.1 | - | 6 | 931 | |||
AK127539.1 | - | 5 | 727 | |||
AK303699.1 | - | 5 | 597 | |||
AK312999.1 | - | 6 | 732 | |||
AY704670.1 | - | 6 | 780 | |||
AY704671.1 | - | 6 | 780 | |||
AY704672.1 | - | 6 | 726 | |||
BC036059.1 | - | 6 | 876 | 5298499 | ||
KU257674.1 | - | 6 | 638 |