Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: LYRM4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001164840.2 | NP_001158312.1 | 2 | 372 | ||
NM_001164841.2 | NP_001158313.1 | 2 | 372 | |||
NM_001318782.1 | NP_001305711.1 | 2 | 372 | |||
NM_001318783.1 | NP_001305712.1 | 2 | 372 | |||
NM_020408.5 | NP_065141.3 | 2 | 372 | |||
NR_104417.1 | 2 | 372 | ||||
NR_134856.1 | 2 | 372 | ||||
XM_017011083.1 | XP_016866572.1 | 2 | 366 | |||
XM_017011084.1 | XP_016866573.1 | 2 | 369 | |||
GenBank mRNA | AF088060.1 | - | 2 | 152 | ||
AF170070.1 | - | 2 | 168 | |||
AF285118.1 | - | 2 | 168 | |||
AK291158.1 | - | 2 | 164 | |||
AK309458.1 | - | 1 | 71 | |||
AK309661.1 | - | 2 | 174 | |||
BC009552.2 | - | 2 | 223 | 3898393 | ||
BI226933.1 | - | 2 | 346 | |||
BI753819.1 | - | 2 | 359 | |||
BI758258.1 | - | 2 | 361 | |||
BX353584.2 | - | 2 | 151 | |||
CK819966.1 | - | 2 | 148 | |||
DA114665.1 | - | 2 | 164 | |||
DA222333.1 | - | 2 | 164 | |||
HY168919.1 | - | 2 | 373 |