Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: RGL3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001035223.3 | NP_001030300.3 | 4 | 472 | ||
NM_001161616.2 | NP_001155088.2 | 4 | 472 | |||
GenBank mRNA | AK057147.1 | - | 4 | 435 | ||
AK293404.1 | - | 4 | 444 | |||
AK298281.1 | - | 3 | 612 | |||
BC014426.1 | - | 4 | 460 | |||
BC064475.1 | - | 3 | 604 | |||
BC101756.1 | - | 4 | 458 | 8069262 | ||
BC111958.1 | - | 4 | 458 | 8327426 | ||
BC143530.1 | - | 4 | 458 | 9052039 | ||
DB184397.1 | - | 4 | 474 |