Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: RNPEPL1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_018226.4 | NP_060696.4 | 7, 8 | 1408 | ||
XM_005247036.4 | XP_005247093.1 | 7, 8 | 1469 | |||
GenBank mRNA | AF300795.1 | - | 7, 8 | 1114 | ||
AK001668.1 | - | 3, 4 | 372 | |||
BC017301.2 | - | 4, 5 | 560 | |||
BC067258.1 | - | 7, 8 | 983 | 6183046 | ||
BC082975.1 | - | 7, 8 | 1308 | 6379537 | ||
BC099906.1 | - | 5, 6 | 1521 | 5311883 |