Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
Integrated CRO-CDMO Services
CDMO Services
CRO Services
Custom Services
Enterprise Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Thermo Fisher Scientific
Search All
Search
Search button
Search button
Close
Order Status
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Services Central
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other RTN4 SIRNA Assays ›
Gene Symbol:
RTN4
Gene Name:
reticulon 4
Gene Aliases:
ASY, NI220/250, NOGO, NOGO-A, NOGOC, NSP, NSP-CL, Nbla00271, Nbla10545, Nogo-B, Nogo-C, RTN-X, RTN4-A, RTN4-B1, RTN4-B2, RTN4-C
Chromosome Location:
Chr.2: 54972187 - 55137831 on Build GRCh38
UniGene ID:
Hs.637850
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID
s32767
Purification
HPLC
Standard
HPLC-IVR
Size
1 nmol
5 nmol
InVivo | 5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
1 nmol
5 nmol
InVivo | 5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
1 nmol
5 nmol
InVivo | 5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
Availability
Inventoried
Catalog #
4392420
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: RTN4
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_001321859.1
NP_001308788.1
6
2920
NM_001321860.1
NP_001308789.1
6
2990
NM_001321861.1
NP_001308790.1
6
2953
NM_001321862.1
NP_001308791.1
6
2940
NM_001321863.1
NP_001308792.1
6
2890
NM_001321904.1
NP_001308833.1
7
3368
NM_007008.2
NP_008939.1
4
610
NM_020532.4
NP_065393.1
6
3673
NM_153828.2
NP_722550.1
4
1216
NM_207520.1
NP_997403.1
5
1273
NM_207521.1
NP_997404.1
6
2931
NR_135829.1
4
496
NR_135830.1
4
474
XM_005264434.3
XP_005264491.1
7
3067
XM_017004518.1
XP_016860007.1
7
3106
XM_017004519.1
XP_016860008.1
7
3223
GenBank mRNA
AB015639.1
-
4
984
AB020693.1
-
6
3509
AB040462.1
-
6
3619
AB040463.1
-
4
1162
AB073351.1
-
4
1345
AF063601.1
-
4
1933
AF077050.1
-
4
609
AF087901.1
-
4
501
AF125103.1
-
4
610
AF132047.1
-
4
1097
AF132048.1
-
4
516
AF148537.1
-
6
3516
AF148538.1
-
4
1059
AF177332.1
-
4
515
AF320999.1
-
5
2679
AF333336.1
-
6
2935
AJ251383.1
-
6
3375
AJ251384.1
-
4
918
AJ251385.1
-
4
396
AK075039.1
-
4
1150
AK090714.1
-
1
76
AK093785.1
-
1
2218
AK129806.1
-
4
1075
AK130812.1
-
4
977
AK172726.1
-
2
40
AY102276.1
-
4
630
AY102277.1
-
4
1162
AY102278.1
-
5
1219
AY102279.1
-
6
3619
AY123245.1
-
6
2932
AY123246.1
-
6
2990
AY123247.1
-
6
2953
AY123248.1
-
6
2939
AY123249.1
-
6
2900
AY123250.1
-
6
2890
BC001035.2
-
4
581
BC007109.1
-
4
507
BC010737.1
-
4
923
BC012619.1
-
4
1073
BC014366.1
-
4
507
BC016165.1
-
5
1222
BC026788.1
-
4
1104
BC068991.1
-
4
1166
BC071848.1
-
5
904
BC150182.1
-
6
3623
40148210
BC152425.1
-
6
3509
8862539
BC152555.1
-
6
3623
40148984
CN429712.1
-
4
445
DQ778739.1
-
4
396
Back To Top
More Information
Other Species Targeted
This siRNA also targets the following transcripts in Mouse: NM_194054, NM_194053, NM_024226, NM_194051, NM_194052, and others.
Gene Ontology Categories:
Function(s)
Process(es)
apoptotic process
axonal fasciculation
cerebral cortex radial glia guided migration
negative regulation of cell growth
negative regulation of axon extension
regulation of apoptotic process
negative regulation of axonogenesis
nuclear pore complex assembly
cardiac epithelial to mesenchymal transition
endoplasmic reticulum tubular network organization
endoplasmic reticulum tubular network assembly
cell-cell adhesion
regulation of branching morphogenesis of a nerve
protein binding
poly(A) RNA binding
cadherin binding involved in cell-cell adhesion
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-7487cff77c-wzvj9:80/10.239.161.253:80.
git-commit: c531a7a49d92b854cb004d8acd820c41186dcba6
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.16.2-2024.09.38-1.0