Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SLC44A2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001145056.1 | NP_001138528.1 | 12 | 1116 | ||
NM_020428.3 | NP_065161.3 | 12 | 1175 | |||
XM_005259997.1 | XP_005260054.1 | 12 | 1175 | |||
XM_005259999.1 | XP_005260056.1 | 12 | 1116 | |||
GenBank mRNA | AK027519.1 | - | 12 | 1060 | ||
AK075512.1 | - | 1 | 77 | |||
AK309801.1 | - | 10 | 945 | |||
AK314582.1 | - | 12 | 1117 | |||
AL832517.1 | - | 2 | 221 | |||
AL832978.1 | - | 9 | 859 | |||
BC010617.2 | - | 9 | 866 | |||
BC040556.1 | - | 12 | 1117 | 5268313 | ||
BC042692.1 | - | 12 | 1117 | |||
FN668568.1 | - | 12 | 1175 |