Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TMEM63C | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_020431.3 | NP_065164.2 | 8 | 735 | ||
GenBank mRNA | AK125159.1 | - | 8 | 667 | ||
BC136613.1 | - | 8 | 617 | 9020603 | ||
BC136614.1 | - | 8 | 614 | 9020604 | ||
BX248756.1 | - | 2 | 111 | |||
BX248759.1 | - | 2 | 82 |