Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TMEM63C | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_020431.3 | NP_065164.2 | 21 | 2154 | ||
GenBank mRNA | AK124749.1 | - | 7 | 662 | ||
AK125159.1 | - | 21 | 2086 | |||
AL157492.1 | - | 2 | 107 | |||
BC136613.1 | - | 21 | 2036 | 9020603 | ||
BC136614.1 | - | 21 | 2033 | 9020604 |