Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PELI1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_020651.3 | NP_065702.2 | 5 | 923 | ||
XM_005264437.3 | XP_005264494.1 | 4 | 742 | |||
XM_011532994.2 | XP_011531296.1 | 5 | 1582 | |||
XM_011532996.2 | XP_011531298.1 | 5 | 2341 | |||
XM_011532997.2 | XP_011531299.1 | 5 | 958 | |||
XM_017004520.1 | XP_016860009.1 | 6 | 1057 | |||
XM_017004521.1 | XP_016860010.1 | 6 | 916 | |||
GenBank mRNA | AF300987.1 | - | 4 | 478 | ||
AF302505.1 | - | 4 | 4501 | |||
AJ278859.1 | - | 4 | 463 | |||
AK027668.1 | - | 2 | 234 | |||
AK312441.1 | - | 5 | 872 | |||
AY364257.1 | - | 4 | 463 | |||
BC050019.1 | - | 5 | 877 | 5296990 | ||
BC050533.1 | - | 5 | 859 | |||
BC063611.1 | - | 5 | 851 | 4992160 | ||
BX649152.1 | - | 1 | 1779 |