Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PELI1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_020651.3 | NP_065702.2 | 4 | 681 | ||
XM_005264437.3 | XP_005264494.1 | 3 | 500 | |||
XM_011532994.2 | XP_011531296.1 | 4 | 1340 | |||
XM_011532996.2 | XP_011531298.1 | 4 | 2099 | |||
XM_011532997.2 | XP_011531299.1 | 3 | 500 | |||
XM_017004520.1 | XP_016860009.1 | 5 | 815 | |||
XM_017004521.1 | XP_016860010.1 | 5 | 674 | |||
GenBank mRNA | AF300987.1 | - | 3 | 236 | ||
AF302505.1 | - | 3 | 4259 | |||
AJ278859.1 | - | 3 | 221 | |||
AK312441.1 | - | 4 | 630 | |||
AY364257.1 | - | 3 | 221 | |||
BC050019.1 | - | 4 | 635 | 5296990 | ||
BC050533.1 | - | 4 | 617 | |||
BC063611.1 | - | 4 | 609 | 4992160 |