Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CORO1B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001018070.2 | NP_001018080.1 | 6 | 599 | ||
NM_020441.2 | NP_065174.1 | 5 | 577 | |||
GenBank mRNA | AK000860.1 | - | 5 | 560 | ||
AK295782.1 | - | 5 | 537 | |||
AK309267.1 | - | 4 | 770 | |||
AK315399.1 | - | 5 | 532 | |||
BC006449.1 | - | 5 | 527 | 3958930 | ||
BI767141.1 | - | 6 | 575 |