Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: COL3A1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_000090.3 | NP_000081.1 | 49 | 4045 | ||
GenBank mRNA | AK091853.1 | - | 2 | 1025 | ||
BC028178.1 | - | 39 | 3134 | 5405119 | ||
X01742.1 | - | 2 | 340 | |||
X06700.1 | - | 10 | 1081 | |||
X14420.1 | - | 49 | 4030 |