Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: NIPAL3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001322854.1 | NP_001309783.1 | 11 | 1524 | ||
NM_001322855.1 | NP_001309784.1 | 12 | 1666 | |||
NM_001322856.1 | NP_001309785.1 | 12 | 1623 | |||
NM_001322857.1 | NP_001309786.1 | 11 | 1444 | |||
NM_001322858.1 | NP_001309787.1 | 12 | 1541 | |||
NM_001322859.1 | NP_001309788.1 | 9 | 1150 | |||
NM_001322860.1 | NP_001309789.1 | 8 | 1090 | |||
NM_001322861.1 | NP_001309790.1 | 8 | 800 | |||
NM_001322862.1 | NP_001309791.1 | 12 | 1501 | |||
NM_001322863.1 | NP_001309792.1 | 11 | 1473 | |||
NM_001322864.1 | NP_001309793.1 | 10 | 1041 | |||
NM_001322866.1 | NP_001309795.1 | 11 | 1364 | |||
NM_001322868.1 | NP_001309797.1 | 9 | 1238 | |||
NM_020448.4 | NP_065181.1 | 11 | 1386 | |||
NR_136506.1 | 10 | 1294 | ||||
NR_136507.1 | 5 | 801 | ||||
NR_136508.1 | 10 | 1255 | ||||
XM_011541808.2 | XP_011540110.1 | 10 | 1301 | |||
XM_017001868.1 | XP_016857357.1 | 10 | 1036 | |||
XM_017001869.1 | XP_016857358.1 | 11 | 1371 | |||
XM_017001870.1 | XP_016857359.1 | 12 | 1636 | |||
GenBank mRNA | AK312433.1 | - | 11 | 1340 | ||
BC063583.1 | - | 11 | 1316 | 4386115 | ||
BX538110.1 | - | 5 | 414 | |||
BX640883.1 | - | 12 | 1448 | |||
U90918.1 | - | 11 | 1277 |