Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: RALGAPA2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_020343.3 | NP_065076.2 | 4 | 433 | ||
XM_005260768.2 | XP_005260825.1 | 4 | 448 | |||
XM_006723598.1 | XP_006723661.1 | 4 | 448 | |||
XM_006723599.1 | XP_006723662.1 | 4 | 448 | |||
XM_011529309.1 | XP_011527611.1 | 4 | 448 | |||
XM_011529310.2 | XP_011527612.1 | 2 | 69 | |||
XM_017027972.1 | XP_016883461.1 | 4 | 448 | |||
XM_017027973.1 | XP_016883462.1 | 2 | 50 | |||
XM_017027974.1 | XP_016883463.1 | 2 | 45 | |||
XM_017027975.1 | XP_016883464.1 | 4 | 352 | |||
XM_017027976.1 | XP_016883465.1 | 2 | 205 | |||
XM_017027977.1 | XP_016883466.1 | 4 | 448 | |||
GenBank mRNA | BC171842.1 | - | 2 | 205 | ||
CR990457.1 | - | 2 | 269 | |||
DQ310704.1 | - | 4 | 290 |