Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: COL4A1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001845.5 | NP_001836.3 | 51, 52 | 5051 | ||
XM_011521048.2 | XP_011519350.1 | 51, 52 | 12983 | |||
GenBank mRNA | AB209646.1 | - | 44, 45 | 3982 | ||
AF258349.1 | - | 4, 5 | 601 | |||
AF363672.1 | - | 4, 5 | 601 | |||
AF400431.1 | - | 4, 5 | 601 | |||
AF536207.1 | - | 4, 5 | 601 | |||
AY285780.1 | - | 4, 5 | 601 | |||
BC047305.1 | - | 26, 27 | 3140 | |||
BC151220.1 | - | 51, 52 | 5035 | 40148649 | ||
DQ464183.1 | - | 4, 5 | 601 | |||
EU260121.1 | - | 4, 5 | 601 | |||
M11315.1 | - | 9, 10 | 1147 |