Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SPRYD7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001127482.2 | NP_001120954.1 | 2 | 405 | ||
NM_020456.3 | NP_065189.1 | 3 | 522 | |||
XM_017020677.1 | XP_016876166.1 | 3 | 522 | |||
GenBank mRNA | AF052154.1 | - | 3 | 321 | ||
AF055016.1 | - | 3 | 321 | |||
AF334405.1 | - | 3 | 361 | |||
AK290576.1 | - | 2 | 244 | |||
AK308818.1 | - | 3 | 355 | |||
AK316576.1 | - | 3 | 328 | |||
BC022519.1 | - | 2 | 211 | 4792366 | ||
DA485722.1 | - | 3 | 523 | |||
DN992366.1 | - | 3 | 518 |