Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: THAP11 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_020457.2 | NP_065190.2 | 1 | 980 | ||
GenBank mRNA | AB015338.1 | - | 1 | 57 | ||
AK289367.1 | - | 1 | 956 | |||
BC012182.1 | - | 1 | 939 | 4554554 | ||
EU869208.1 | - | 1 | 349 | |||
L10378.1 | - | 1 | 817 | |||
L10379.1 | - | 1 | 817 |