Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PPP4R3B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001122964.2 | NP_001116436.2 | 4 | 1067 | ||
NM_001282850.1 | NP_001269779.1 | 4 | 1067 | |||
NM_020463.3 | NP_065196.1 | 4 | 1067 | |||
XM_005264442.2 | XP_005264499.1 | 4 | 1067 | |||
XM_005264444.2 | XP_005264501.1 | 4 | 1067 | |||
XM_005264445.2 | XP_005264502.1 | 4 | 1067 | |||
XM_005264446.2 | XP_005264503.1 | 4 | 1067 | |||
XM_017004531.1 | XP_016860020.1 | 4 | 1067 | |||
XM_017004537.1 | XP_016860026.1 | 4 | 1067 | |||
XM_017004538.1 | XP_016860027.1 | 4 | 1099 | |||
XM_017004539.1 | XP_016860028.1 | 4 | 959 | |||
XM_017004540.1 | XP_016860029.1 | 4 | 1099 | |||
GenBank mRNA | AB037808.1 | - | 4 | 930 | ||
AK026437.1 | - | 1 | 345 | |||
AK056036.1 | - | 4 | 964 | |||
AY825268.1 | - | 4 | 627 | |||
AY825269.1 | - | 4 | 627 | |||
BC006215.2 | - | 4 | 938 | 3140319 | ||
BC060855.1 | - | 4 | 956 | 5259789 |