Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SMAGP | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001031628.1 | NP_001026798.1 | 4 | 376 | ||
NM_001033873.1 | NP_001029045.1 | 4 | 365 | |||
GenBank mRNA | AF091087.1 | - | 1 | 55 | ||
AK001839.1 | - | 4 | 339 | |||
BC001400.2 | - | 4 | 344 | |||
BC003379.1 | - | 1 | 51 | |||
BC008712.1 | - | 4 | 353 | 3546069 | ||
BC045658.1 | - | 4 | 848 | |||
BC120965.1 | - | 4 | 341 | 40115343 | ||
BC120966.1 | - | 4 | 341 | 40115344 | ||
BG740791.1 | - | 4 | 385 | |||
CA776511.1 | - | 4 | 397 |