Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TMEM27 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_020665.5 | NP_065716.1 | 5 | 719 | ||
XM_017029680.1 | XP_016885169.1 | 5 | 587 | |||
GenBank mRNA | AF229179.1 | - | 5 | 486 | ||
AK313835.1 | - | 5 | 544 | |||
AY359060.1 | - | 5 | 469 | |||
BC014317.1 | - | 5 | 723 | 4048217 | ||
BC015099.1 | - | 5 | 551 | 3829035 | ||
BC050606.1 | - | 5 | 471 | 5183554 |