Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: COL4A5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_000495.4 | NP_000486.1 | 13 | 1015 | ||
NM_033380.2 | NP_203699.1 | 13 | 1015 | |||
XM_011530849.2 | XP_011529151.2 | 14 | 763 | |||
XM_017029259.1 | XP_016884748.1 | 14 | 763 | |||
XM_017029260.1 | XP_016884749.1 | 14 | 763 | |||
XM_017029261.1 | XP_016884750.1 | 14 | 763 | |||
XM_017029262.1 | XP_016884751.1 | 14 | 763 | |||
GenBank mRNA | BC151846.1 | - | 13 | 977 | 8860442 | |
KM282618.1 | - | 13 | 1015 | |||
M58526.1 | - | 10 | 491 | |||
M90464.1 | - | 13 | 935 |