Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MIF4GD | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001242498.1 | NP_001229427.1 | 3 | 269 | ||
NM_001242500.1 | NP_001229429.1 | 2 | 171 | |||
NM_020679.3 | NP_065730.2 | 3 | 248 | |||
XM_005257530.1 | XP_005257587.1 | 3 | 390 | |||
XM_011525051.2 | XP_011523353.1 | 3 | 485 | |||
XM_011525052.2 | XP_011523354.1 | 3 | 464 | |||
XM_011525054.2 | XP_011523356.1 | 3 | 485 | |||
XM_017024888.1 | XP_016880377.1 | 3 | 485 | |||
XM_017024889.1 | XP_016880378.1 | 3 | 269 | |||
XM_017024890.1 | XP_016880379.1 | 3 | 295 | |||
XM_017024891.1 | XP_016880380.1 | 3 | 207 | |||
XM_017024895.1 | XP_016880384.1 | 3 | 248 | |||
GenBank mRNA | AK300711.1 | - | 3 | 208 | ||
BC033759.1 | - | 2 | 147 | 5215495 | ||
BU190758.1 | - | 2 | 170 | |||
BX404175.2 | - | 3 | 191 | |||
KU178731.1 | - | 2 | 111 | |||
KU178732.1 | - | 2 | 132 |