Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: AS3MT | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_020682.3 | NP_065733.2 | 8 | 770 | ||
GenBank mRNA | AF226730.1 | - | 8 | 707 | ||
AK057833.1 | - | 8 | 789 | |||
AK225972.1 | - | 8 | 789 | |||
AK309737.1 | - | 8 | 702 | |||
BC119637.1 | - | 8 | 674 | |||
BC119638.2 | - | 8 | 674 |
Gene Symbol: BORCS7-ASMT | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_037644.1 | 12 | 1035 |