Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SLC24A3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_020689.3 | NP_065740.2 | 13 | 1574 | ||
GenBank mRNA | AF169257.2 | - | 13 | 1320 | ||
AF288087.1 | - | 13 | 1574 |