Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
Integrated CRO-CDMO Services
CDMO Services
CRO Services
Custom Services
Enterprise Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Thermo Fisher Scientific
Search All
Search
Search button
Search button
Close
Order Status
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Services Central
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other NDRG2 SIRNA Assays ›
Gene Symbol:
NDRG2
Gene Name:
NDRG family member 2
Gene Aliases:
SYLD
Chromosome Location:
Chr.14: 21016763 - 21070872 on Build GRCh38
UniGene ID:
Hs.525205
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID
s33036
Purification
HPLC
Standard
HPLC-IVR
Size
1 nmol
5 nmol
InVivo | 5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
1 nmol
5 nmol
InVivo | 5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
1 nmol
5 nmol
InVivo | 5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
Availability
Inventoried
Catalog #
4392420
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: NDRG2
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_001282211.1
NP_001269140.1
11, 12
828
NM_001282213.1
NP_001269142.1
12, 13
1107
NM_001282214.1
NP_001269143.1
12, 13
1095
NM_001282216.1
NP_001269145.1
11, 12
843
NM_001320329.1
NP_001307258.1
12, 13
1760
NM_016250.2
NP_057334.1
12, 13
950
NM_201535.1
NP_963293.1
13, 14
992
NM_201536.1
NP_963294.1
11, 12
883
NM_201537.1
NP_963831.1
12, 13
925
NM_201538.1
NP_963832.1
11, 12
860
NM_201539.1
NP_963833.1
12, 13
902
NM_201540.1
NP_963834.1
13, 14
969
NM_201541.1
NP_963835.1
12, 13
927
XM_011536996.2
XP_011535298.1
13, 14
979
XM_011536997.1
XP_011535299.1
13, 14
980
XM_011536998.1
XP_011535300.1
13, 14
970
XM_011536999.1
XP_011535301.1
11, 12
1375
XM_011537001.1
XP_011535303.1
12, 13
933
XM_011537002.1
XP_011535304.1
12, 13
1079
GenBank mRNA
AB033074.1
-
11, 12
812
AF087872.1
-
11, 12
866
AF159092.3
-
12, 13
873
AF304051.1
-
13, 14
992
AK024521.1
-
13, 14
925
AK057843.1
-
12, 13
896
AK057847.1
-
11, 12
860
AK090657.1
-
11, 12
799
AK096999.1
-
12, 13
1760
AK098388.1
-
920
AK130029.1
-
11, 12
798
AK293514.1
-
11, 12
828
AL136574.1
-
11, 12
837
BC010458.1
-
11, 12
831
4215141
BC093038.1
-
12, 13
926
4811718
BX248031.1
-
9, 10
702
BX647091.1
-
9, 10
1025
BX647748.1
-
12, 13
1042
CR749252.1
-
12, 13
1095
Back To Top
More Information
Panther Classification:
Molecular Function -
serine protease
Gene Ontology Categories:
Function(s)
Process(es)
negative regulation of cytokine production
signal transduction
regulation of vascular endothelial growth factor production
Wnt signaling pathway
substantia nigra development
cell differentiation
negative regulation of smooth muscle cell proliferation
negative regulation of ERK1 and ERK2 cascade
regulation of platelet-derived growth factor production
molecular_function
protein binding
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-green-cccc65c97-6lbnp:80/10.239.161.162:80.
git-commit: 2b810224bbfb944e11adb8d8c7a69f36ca15064f
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.16.1-offline