Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: BIRC6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_016252.3 | NP_057336.3 | 3 | 682 | ||
XM_005264449.4 | XP_005264506.2 | 3 | 803 | |||
XM_005264450.4 | XP_005264507.2 | 3 | 803 | |||
XM_005264451.4 | XP_005264508.2 | 3 | 803 | |||
XM_005264452.4 | XP_005264509.2 | 3 | 548 | |||
XM_005264453.4 | XP_005264510.2 | 3 | 548 | |||
XM_005264454.4 | XP_005264511.2 | 3 | 803 | |||
XM_005264455.4 | XP_005264512.2 | 3 | 803 | |||
XM_006712054.3 | XP_006712117.1 | 3 | 803 | |||
XM_006712055.3 | XP_006712118.1 | 3 | 803 | |||
XM_006712056.3 | XP_006712119.1 | 3 | 803 | |||
XM_011533003.2 | XP_011531305.1 | 3 | 248 | |||
XM_011533005.2 | XP_011531307.1 | 3 | 803 | |||
XM_017004556.1 | XP_016860045.1 | 3 | 803 | |||
XM_017004557.1 | XP_016860046.1 | 3 | 803 | |||
XM_017004558.1 | XP_016860047.1 | 3 | 803 | |||
XM_017004559.1 | XP_016860048.1 | 3 | 803 | |||
XM_017004560.1 | XP_016860049.1 | 3 | 803 | |||
GenBank mRNA | AF265555.1 | - | 3 | 464 | ||
BX345326.2 | - | 3 | 682 |