Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: COL4A6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001287758.1 | NP_001274687.1 | 45 | 4900 | ||
NM_001287759.1 | NP_001274688.1 | 43 | 4777 | |||
NM_001287760.1 | NP_001274689.1 | 42 | 4678 | |||
NM_001847.3 | NP_001838.2 | 44 | 4744 | |||
NM_033641.3 | NP_378667.1 | 44 | 4849 | |||
XM_006724617.3 | XP_006724680.1 | 45 | 4789 | |||
XM_011530852.2 | XP_011529154.1 | 44 | 4717 | |||
XM_011530853.2 | XP_011529155.1 | 43 | 5505 | |||
GenBank mRNA | AK300502.1 | - | 3 | 410 | ||
AK302734.1 | - | 1 | 311 | |||
BC140855.1 | - | 42 | 4574 | 9021723 | ||
BC144669.1 | - | 43 | 4673 | 9053200 | ||
D21337.1 | - | 43 | 4813 | |||
DA748513.1 | - | 2 | 227 | |||
EU182215.1 | - | 2 | 232 | |||
U04845.1 | - | 44 | 4727 |