Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TENM2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001080428.2 | NP_001073897.2 | 25 | 7391 | ||
NM_001122679.1 | NP_001116151.1 | 29 | 7963 | |||
XM_005265950.2 | XP_005266007.1 | 31 | 8610 | |||
XM_005265952.2 | XP_005266009.1 | 27 | 7337 | |||
XM_006714897.2 | XP_006714960.1 | 30 | 8589 | |||
XM_011534604.2 | XP_011532906.1 | 30 | 8424 | |||
XM_017009660.1 | XP_016865149.1 | 31 | 8637 | |||
XM_017009661.1 | XP_016865150.1 | 30 | 8616 | |||
XM_017009662.1 | XP_016865151.1 | 30 | 8427 | |||
XM_017009663.1 | XP_016865152.1 | 28 | 7832 | |||
XM_017009664.1 | XP_016865153.1 | 28 | 7745 | |||
XM_017009665.1 | XP_016865154.1 | 28 | 7541 | |||
XM_017009666.1 | XP_016865155.1 | 27 | 7520 | |||
XM_017009667.1 | XP_016865156.1 | 28 | 7514 | |||
XM_017009668.1 | XP_016865157.1 | 28 | 9051 | |||
XM_017009669.1 | XP_016865158.1 | 28 | 8883 | |||
GenBank mRNA | AB032953.2 | - | 20 | 6100 | ||
AK293262.1 | - | 3 | 822 | |||
AK302302.1 | - | 3 | 1528 | |||
AL137500.1 | - | 4 | 2216 | |||
BX648178.1 | - | 24 | 6985 | |||
CR936703.1 | - | 25 | 7324 |