Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: REXO1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_020695.3 | NP_065746.3 | 15, 16 | 3672 | ||
XM_017027029.1 | XP_016882518.1 | 15, 16 | 3734 | |||
GenBank mRNA | AB032964.1 | - | 15, 16 | 3660 | ||
AK301729.1 | - | 11, 12 | 1709 | |||
AK302362.1 | - | 12, 13 | 1616 | |||
BC032244.1 | - | 15, 16 | 3682 | 4398472 |