Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: KIAA1143 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001320334.1 | NP_001307263.1 | 3 | 419 | ||
NM_020696.3 | NP_065747.1 | 3 | 364 | |||
GenBank mRNA | AB032969.1 | - | 2 | 171 | ||
AK289549.1 | - | 3 | 336 | |||
BC008468.1 | - | 3 | 339 | 4277104 | ||
BC016790.1 | - | 3 | 298 | 4102473 | ||
BC070360.1 | - | 3 | 294 | |||
CD049061.1 | - | 3 | 361 | |||
DA183520.1 | - | 3 | 357 | |||
DA192476.1 | - | 3 | 339 | |||
DB238094.1 | - | 3 | 375 |