Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: AMIGO1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_020703.2 | NP_065754.2 | 2 | 1168 | ||
XM_011541812.2 | XP_011540114.1 | 2 | 1174 | |||
GenBank mRNA | AB032989.1 | - | 1 | 620 | ||
AK295678.1 | - | 2 | 1147 | |||
AY237007.1 | - | 1 | 786 | |||
BC040879.1 | - | 2 | 1136 | 5725643 |