Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: COL5A2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_000393.3 | NP_000384.2 | 53, 54 | 4625 | ||
XM_011510573.2 | XP_011508875.1 | 53, 54 | 4407 | |||
GenBank mRNA | AB209045.1 | - | 53, 54 | 4487 | ||
AK300146.1 | - | 5, 6 | 1102 | |||
BC043613.1 | - | 15, 16 | 1720 | |||
BC086874.1 | - | 37, 38 | 3614 | 30708812 | ||
M11718.1 | - | 3, 4 | 663 | |||
X04758.1 | - | 35, 36 | 3150 | |||
Y14690.1 | - | 53, 54 | 4479 |