Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CNOT6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001303241.1 | NP_001290170.1 | 12 | 1574 | ||
XM_005265953.1 | XP_005266010.1 | 11 | 1574 | |||
XM_011534605.2 | XP_011532907.1 | 10 | 1467 | |||
XM_011534606.1 | XP_011532908.1 | 10 | 1482 | |||
XM_011534607.1 | XP_011532909.1 | 11 | 1677 | |||
XM_017009670.1 | XP_016865159.1 | 9 | 1359 | |||
XM_017009671.1 | XP_016865160.1 | 9 | 1021 | |||
XM_017009672.1 | XP_016865161.1 | 10 | 1482 | |||
GenBank mRNA | AB033020.1 | - | 12 | 1540 | ||
BC152469.1 | - | 12 | 1540 | 8862606 |