Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ZNF512B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_020713.2 | NP_065764.1 | 5 | 997 | ||
XM_005260226.3 | XP_005260283.1 | 6 | 1069 | |||
XM_011528929.2 | XP_011527231.1 | 5 | 959 | |||
XM_011528930.2 | XP_011527232.1 | 5 | 1186 | |||
XM_011528933.2 | XP_011527235.1 | 5 | 1182 | |||
GenBank mRNA | AB033022.1 | - | 3 | 816 | ||
BC125128.1 | - | 5 | 988 | 40119047 | ||
BC125129.1 | - | 5 | 987 | 40119053 |