Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ZNF564 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AL834146.1 | - | 1 | 326 |
Gene Symbol: ZNF490 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_020714.2 | NP_065765.1 | 5 | 1554 | ||
GenBank mRNA | AB033024.1 | - | 5 | 1548 | ||
BC038586.1 | - | 5 | 1537 | 5587423 |