Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PLEKHH1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_020715.2 | NP_065766.1 | 24 | 3521 | ||
XM_011537009.1 | XP_011535311.1 | 24 | 3697 | |||
XM_011537015.2 | XP_011535317.1 | 17 | 3752 | |||
XM_017021499.1 | XP_016876988.1 | 23 | 3678 | |||
XM_017021500.1 | XP_016876989.1 | 24 | 3613 | |||
XM_017021501.1 | XP_016876990.1 | 23 | 3594 | |||
XM_017021502.1 | XP_016876991.1 | 17 | 3474 | |||
XM_017021503.1 | XP_016876992.1 | 17 | 2834 | |||
XM_017021504.1 | XP_016876993.1 | 19 | 2976 | |||
GenBank mRNA | AB033026.1 | - | 24 | 3497 | ||
AK124787.1 | - | 1 | 578 | |||
AK293950.1 | - | 5 | 562 | |||
AK295896.1 | - | 17 | 2221 | |||
BC146787.1 | - | 24 | 3497 | 9007246 | ||
BX648137.1 | - | 2 | 1878 | |||
BX648464.1 | - | 15 | 2633 |