Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: COL6A1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001848.2 | NP_001839.2 | 5, 6 | 817 | ||
GenBank mRNA | AK298968.1 | - | 5, 6 | 796 | ||
AK299396.1 | - | 5, 6 | 799 | |||
AK307989.1 | - | 5, 6 | 796 | |||
BC052575.1 | - | 5, 6 | 797 | 6598940 | ||
X15879.1 | - | 5, 6 | 763 |