Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: USP31 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_020718.3 | NP_065769.3 | 9 | 1564 | ||
XM_017023496.1 | XP_016878985.1 | 9 | 1595 | |||
XM_017023497.1 | XP_016878986.1 | 9 | 3090 | |||
XM_017023498.1 | XP_016878987.1 | 8 | 877 | |||
XM_017023499.1 | XP_016878988.1 | 6 | 681 | |||
XM_017023500.1 | XP_016878989.1 | 9 | 1595 | |||
GenBank mRNA | AJ586135.2 | - | 9 | 1564 | ||
AK126752.1 | - | 9 | 1248 | |||
BM842261.1 | - | 3 | 261 |